missing translation for 'onlineSavingsMsg'
Learn More

Taurine, 99.6%, MP Biomedicals™

$67.01 - $67.01

Chemical Identifiers

CAS 107-35-7
Molecular Formula C2H7NO3S
Molecular Weight (g/mol) 125.14
MDL Number MFCD00008197
InChI Key XOAAWQZATWQOTB-UHFFFAOYSA-N
Synonym tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
PubChem CID 70681
ChEBI CHEBI:51988
IUPAC Name 2-aminoethane-1-sulfonic acid
SMILES NCCS(O)(=O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
ICN10300310
View Documents
MP Biomedicals Inc
0210300310
10 g
Each for $67.01
Only null left
 
Description

Description

Specifications

Chemical Identifiers

107-35-7
125.14
XOAAWQZATWQOTB-UHFFFAOYSA-N
70681
2-aminoethane-1-sulfonic acid
C2H7NO3S
MFCD00008197
tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
CHEBI:51988
NCCS(O)(=O)=O

Specifications

107-35-7
≥99%
MFCD00008197
tetrabutylammonium chloride, tetrabutyl ammonium chloride, tetra-n-butylammonium chloride, tbac, tetrabutylazanium chloride, 1-butanaminium, n,n,n-tributyl-, chloride, tetrabutylammoniumchloride, 1-butanaminium, n,n,n-tributyl-, chloride 1:1, n,n,n-tributyl-1-butanaminium chloride, n,n,n-tributylbutan-1-aminium chloride
NCCS(O)(=O)=O
125.14
CHEBI:51988
0.996
Taurine
White
C2H7NO3S
10 g
XOAAWQZATWQOTB-UHFFFAOYSA-N
2-aminoethane-1-sulfonic acid
70681
125.15
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 203-483-8

Recommended Storage : Store at Room Temperature (15° to 30°C)

SDS
missing translation for 'documents'

missing translation for 'documents'