Learn More
SignalChem Biotech Tau-441, DYRK1A-phosphorylated & Biotinylated 100ug
Catalog No. 502572875
Encompass
Recombinant human tag-free Tau-441 was co-expressed with DYRK1A in E. coli cells, phosphorylated by DYRK1A in vivo and in vitro, then the purified protein was chemically biotinylated in vitro.
Catalog No. 502572875 Supplier SignalChem Biotech Supplier No. T0850RNB100
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human tag-free Tau-441 was co-expressed with DYRK1A in E. coli cells, phosphorylated by DYRK1A in vivo and in vitro, then the purified protein was chemically biotinylated in vitro.