Recombinant human Tau-441 (P301L) expressed in E. coli cells with an N-terminal Avi tag and enzymatically biotinylated in vitro prior to the final chromatography purification.
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Recombinant human Tau-441 (P301L) expressed in E. coli cells with an N-terminal Avi tag and enzymatically biotinylated in vitro prior to the final chromatography purification.