missing translation for 'onlineSavingsMsg'
Learn More

TATU 98.0+%, TCI America™

Catalog No. A18611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18611G 1 g
A18615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18611G Supplier TCI America Supplier No. A18611G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 873798-09-5
Molecular Formula C10H15BF4N6O
Molecular Weight (g/mol) 322.075
InChI Key AUPDFAPCZZXFMX-UHFFFAOYSA-N
Synonym 1-[Bis(dimethylamino)methylene]-1H-1,2,3-triazolo[4,5-b]pyridinium 3-Oxide Tetrafluoroborate
PubChem CID 11099301
IUPAC Name [dimethylamino(triazolo[4,5-b]pyridin-3-yloxy)methylidene]-dimethylazanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.CN(C)C(=[N+](C)C)ON1C2=C(C=CC=N2)N=N1

Specifications

Melting Point 184°C
Color White-Yellow
Quantity 1 g
UN Number 1759
Formula Weight 322.07
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material TATU
TSCA No
Recommended Storage Refrigerator