missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TAT-Gap19
Description
TAT-Gap19 is a Cx43 hemichannel blocker (IC50 ~7 μM). No significant affinity for gap junctions or Panx1 channels. N-terminal transactivator of transcription (TAT) motif promotes membrane permeability and increases inhibitory effect of Gap19 (Cat. No. 5353). Active in vivo. Brain penetrant.
Specifications
Specifications
| Purity | 95% |
| CAS | 1507930-54-2 |
| Content And Storage | Store at −20°C |
| Molecular Weight (g/mol) | 2703.28g/mol |
| Quantity | 1 mg |
| Regulatory Status | RUO - research use only |
| Sequence | YGRKKRRQRRRKQIEIKKFK |
| Specific Reactivity | Cx43 hemichannel blocker (IC50=7μM) |
For Research Use Only