missing translation for 'onlineSavingsMsg'
Learn More

TAR 99.0+%, TCI America™

Catalog No. A50021G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A50021G 1 g
A50025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A50021G Supplier TCI America Supplier No. A50021G
Only null left

[Metal indicator and spectrophotometric reagent for transition metals]

Chemical Identifiers

CAS 2246-46-0
Molecular Formula C9H7N3O2S
Molecular Weight (g/mol) 221.23
MDL Number MFCD00005322
InChI Key UVQWEDPLIFNWSH-YRNVUSSQSA-N
Synonym 4-2-thiazolylazo resorcinol, 1,3-benzenediol, 4-2-thiazolylazo, 2-2,4-dihydroxyphenylazo thiazole, 4-thiazol-2-ylazo resorcinol, 4-thiazol-2-yldiazenyl benzene-1,3-diol, 4-2-1,3-thiazol-2-yl diazen-1-yl benzene-1,3-diol, 4-e-2-1,3-thiazol-2-yl diazen-1-yl benzene-1,3-diol, shnikuxmzfppcs-vawyxsnfsa-n, 4-e-1,3-thiazol-2-yldiazenyl-1,3-benzenediol #
PubChem CID 9571124
IUPAC Name (4E)-3-hydroxy-4-[2-(1,3-thiazol-2-yl)hydrazin-1-ylidene]cyclohexa-2,5-dien-1-one
SMILES OC1=CC(=O)C=C\C1=N/NC1=NC=CS1

Specifications

Melting Point 209°C
Quantity 1 g
Formula Weight 221.23
Percent Purity ≥99.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material TAR [=4-(2-Thiazolylazo)resorcinol] [Metal indicator and spectrophotometric reagent for trans