missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ SARS-CoV-2 Control Dilution Buffer

Catalog No. A49889
Encompass
Change view
Click to view available options
Quantity:
10 x 250 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A49889 10 x 250 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A49889 Supplier Applied Biosystems™ Supplier No. A49889

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Applied Biosystems TaqMan SARS-CoV-2 Control Dilution Buffer is used to dilute the TaqMan SARS-CoV-2 RNA Control (Cat. No. 956118), which is used with the TaqMan SARS-CoV-2 Pooling Assay Kit (Cat. No. A50790).

The TaqMan SARS-CoV-2 Control Dilution Buffer is also used to dilute the TaqMan SARS-CoV-2, Flu A, Flu B RNA Control and TaqMan SARS-CoV-2, Flu A/B, RSV RNA Control, which are used with the TaqMan SARS-CoV-2, Flu A, Flu B RT-PCR Assay Kit (Cat. No. A47701) and TaqMan SARS-CoV-2, Flu A/B, RSV RT-PCR Assay Kit (Cat. No. A47702).

Specifications

Content And Storage TaqMan SARS CoV-2 Control Dilution Buffer, 1 x 250 μL/kit, 10 kits included

Store at -10 to -30°C.
Detection Method Primer-Probe Detection
For Use With (Application) SARS CoV-2 Pathogen Research Solutions
Product Line TaqMan
Quantity 10 x 250 μL
Shipping Condition Dry Ice
Species Human
Target Organism Class severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)

For Research Use Only. Not for use in diagnostic procedures.