missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq Pol Bulk

Catalog No. p-8015249
Change view
Click to view available options
Quantity:
10 x 250U
1000U
250U
3 x 250U
5 x 1000U
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1EPTQA025 250U
11EPTQA325 3 x 250U
11EPTQA100 1000U
11EPTQA925 10 x 250U
11EPTQA105 5 x 1000U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MP1EPTQA025 Supplier MP Biomedicals™ Supplier No. 11EPTQA025
Only null left

For life science applications

  • Ready for CE IVD labeling of clinical applications

Specifications

Concentration 5 U/μL
Description Taq Pol Bulk
Quantity 250U
Includes Taq DNA Polymerase, 10X Standard Buffer with Mg, 10X Standard Buffer without Mg, 25mM MgCl2

For Resaerch Use Only