Learn More
New England Biolab Ltd Taq DNA Polymerase with Standard Taq Buffer 400 units
Catalog No. 50995269
Encompass
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. It is supplied with 10X Standard Taq Reaction Buffer, which is detergent-free and designed to be compatible with existing assay systems.
Catalog No. 50995269 Supplier New England Biolab Ltd Supplier No. M0273S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Taq DNA Polymerase is a thermostable DNA polymerase that possesses a 5' to 3' polymerase activity and a 5' flap endonuclease activity. It is supplied with 10X Standard Taq Reaction Buffer, which is detergent-free and designed to be compatible with existing assay systems.