missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Taq CORE Kit 10

Catalog No. MP1EPTQS901
Change view
Click to view available options
Quantity:
10 × 250 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP1EPTQS901 10 × 250 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP1EPTQS901 Supplier MP Biomedicals™ Supplier No. 11EPTQS901K

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Designed for the amplification of specific regions of DNA using PCR. MP Biomedicals Taq CORE Kit 10 provides the enzymes, buffers and dNTPs necessary for amplification, including the (NH4)2SO4 buffer, which has been report

Specifications

Concentration 5 U/μL
Content And Storage Freeze at -20°C
Form Frozen
Quantity 10 × 250 units
For Use With (Application) Standard PCR