missing translation for 'onlineSavingsMsg'
Learn More

TAPS, 99%

CAS: 29915-38-6 | C7H17NO6S | 243.27 g/mol

$85.97 - $206.53

Chemical Identifiers

CAS 29915-38-6
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.27
MDL Number MFCD00007538
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
Synonym taps, 3-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino propane-1-sulfonic acid, unii-y5dc3in066, n-tris hydroxymethyl methyl-3-aminopropanesulfonic acid, 2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, taps buffer, 1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino
PubChem CID 121591
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1775414
View Documents
Thermo Scientific Chemicals
A1775414
25 g
Each for $85.97
Only null left
 
AAA1775422
View Documents
Thermo Scientific Chemicals
A1775422
100 g
Each for $206.53
Only null left
 
Description

Description

A zwitterionic Good's Buffer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

29915-38-6
243.27
YNLCVAQJIKOXER-UHFFFAOYSA-N
121591
OCC(CO)(CO)NCCCS(O)(=O)=O
C7H17NO6S
MFCD00007538
taps, 3-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino propane-1-sulfonic acid, unii-y5dc3in066, n-tris hydroxymethyl methyl-3-aminopropanesulfonic acid, 2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, taps buffer, 1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid

Specifications

29915-38-6
C7H17NO6S
25 g
YNLCVAQJIKOXER-UHFFFAOYSA-N
3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
121591
99%
239°C to 241°C (decomposition)
MFCD00007538
taps, 3-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino propane-1-sulfonic acid, unii-y5dc3in066, n-tris hydroxymethyl methyl-3-aminopropanesulfonic acid, 2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, taps buffer, 1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino
OCC(CO)(CO)NCCCS(O)(=O)=O
243.27
243.28
TAPS
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 249-954-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only