missing translation for 'onlineSavingsMsg'
Learn More

TAPS, 99%

Catalog No. AAA1775414
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1775414 25 g
AAA1775422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1775414 Supplier Thermo Scientific Chemicals Supplier No. A1775414

CAS: 29915-38-6 | C7H17NO6S | 243.27 g/mol

A zwitterionic Good's Buffer

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 29915-38-6
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.27
MDL Number MFCD00007538
InChI Key YNLCVAQJIKOXER-UHFFFAOYSA-N
Synonym taps, 3-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino propane-1-sulfonic acid, unii-y5dc3in066, n-tris hydroxymethyl methyl-3-aminopropanesulfonic acid, 2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino-1-propanesulfonic acid, taps buffer, 1-propanesulfonic acid, 3-2-hydroxy-1,1-bis hydroxymethyl ethyl amino
PubChem CID 121591
IUPAC Name 3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonic acid
SMILES OCC(CO)(CO)NCCCS(O)(=O)=O

Specifications

Melting Point 239°C to 241°C (decomposition)
Quantity 25 g
Formula Weight 243.28
Percent Purity 99%
Chemical Name or Material TAPS
EINECSNumber 249-954-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only