missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum wire, 1.5mm (0.06in) dia, annealed, 99.95% (metals basis)
CAS: 7440-25-7 | Ta | 180.95 g/mol
Supplier: Thermo Scientific Chemicals 042933CC
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Tantalum wire | |
| 7440-25-7 | |
| Odorless | |
| Wire | |
| Ta | |
| tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
| [Ta] | |
| 180.95 | |
| CHEBI:33348 |
| 1.5mm (0.06 in.) dia., Annealed, Temper: soft | |
| 500 cm | |
| ≈29.3g/m | |
| (metals basis) | |
| MFCD00011252 | |
| GUVRBAGPIYLISA-UHFFFAOYSA-N | |
| tantalum | |
| 23956 | |
| 99.95% |
Chemical Identifiers
| 7440-25-7 | |
| 180.95 | |
| GUVRBAGPIYLISA-UHFFFAOYSA-N | |
| 23956 | |
| tantalum |
| Ta | |
| MFCD00011252 | |
| tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
| CHEBI:33348 | |
| [Ta] |
Safety and Handling
EINECSNumber : 231-135-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only