missing translation for 'onlineSavingsMsg'
Learn More

Tantalum wire, 0.203mm (0.008in) dia, hard, 99.9% (metals basis)

Catalog No. AA45133G1
Change view
Click to view available options
Quantity:
1 m
10 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45133G1 1 m
AA45133G6 10 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA45133G1 Supplier Thermo Scientific Chemicals Supplier No. 045133G1

CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Melting Point 2996°C
Boiling Point 5429°C
Density 16.69
Quantity 1 m
Packaging Tube
Formula Weight 180.95
Percent Purity 99.9%
Chemical Name or Material Tantalum wire
EINECSNumber 231-135-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only