missing translation for 'onlineSavingsMsg'
Learn More

Tantalum wire, 0.025mm (0.001in) dia, hard, 99.9% (metals basis)

Catalog No. p-4782172
Change view
Click to view available options
Quantity:
1 m
10 m
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40675G1 1 m
AA40675G6 10 m
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA40675G1 Supplier Thermo Scientific Chemicals Supplier No. 040675G1
Only null left

Form: Approx. 0.0082g/m; Temper: as drawn | CAS: 7440-25-7 | Ta | 180.95 g/mol

Microelectronics, contacts, surgical, high temperature

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Color Gray to Silver
Appearance Gray to silver wire
Density 16.69g/cm3
Quantity 1 m
Odor Odorless
Form Wire
Assay Percent Range 99.9% (Metals basis)
Chemical Name or Material Tantalum, Hard
EINECSNumber 231-135-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only