missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum(V) Chloride Anhydrous 95.0+%, TCI America™
$106.25 - $328.08
Chemical Identifiers
| CAS | 7721-01-9 |
|---|---|
| Molecular Formula | Cl5Ta |
| Molecular Weight (g/mol) | 358.20 |
| MDL Number | MFCD00011253 |
| InChI Key | OEIMLTQPLAGXMX-UHFFFAOYSA-I |
| IUPAC Name | tantalum(5+) pentachloride |
| SMILES | [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5] |
Chemical Identifiers
| 7721-01-9 | |
| 358.20 | |
| OEIMLTQPLAGXMX-UHFFFAOYSA-I | |
| [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5] |
| Cl5Ta | |
| MFCD00011253 | |
| tantalum(5+) pentachloride |
Specifications
| 239°C | |
| White | |
| Crystalline Powder | |
| Cl5Ta | |
| 3260 | |
| [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5] | |
| 358.20 | |
| ≥95.0% (T) |
| 220°C | |
| 7721-01-9 | |
| 5 g | |
| MFCD00011253 | |
| OEIMLTQPLAGXMX-UHFFFAOYSA-I | |
| tantalum(5+) pentachloride | |
| 358.20 | |
| Tantalum(V) Chloride Anhydrous |
Safety and Handling
EINECSNumber : (1)-0654
RTECSNumber : WW5600000
TSCA : Yes