missing translation for 'onlineSavingsMsg'
Learn More

Tantalum(V) Chloride Anhydrous 95.0+%, TCI America™

Catalog No. T26845G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T26845G 5 g
T268425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T26845G Supplier TCI America Supplier No. T26845G
Only null left

Chemical Identifiers

CAS 7721-01-9
Molecular Formula Cl5Ta
Molecular Weight (g/mol) 358.20
MDL Number MFCD00011253
InChI Key OEIMLTQPLAGXMX-UHFFFAOYSA-I
IUPAC Name tantalum(5+) pentachloride
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ta+5]

Specifications

Boiling Point 239°C
Melting Point 220°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
UN Number 3260
Formula Weight 358.20
Percent Purity ≥95.0% (T)
Chemical Name or Material Tantalum(V) Chloride Anhydrous
EINECSNumber (1)-0654
RTECSNumber WW5600000
TSCA Yes