missing translation for 'onlineSavingsMsg'
Learn More

Tantalum Thinfoil, 0.0127mm (0.0005in) thick, 99.95% (metals basis)

Catalog No. p-4408175
Change view
Click to view available options
Quantity:
50 x 100 mm
50 x 500 mm
50 x 2000 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA14266FY 50 x 100 mm
AA14266GD 50 x 500 mm
AA142668S 50 x 2000 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA14266FY Supplier Thermo Scientific Chemicals Supplier No. 014266FY
Only null left

CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Quantity 50 x 100 mm
Assay Percent Range 99.95% (metals basis)
Chemical Name or Material Tantalum Thinfoil
EINECSNumber 231-135-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only