missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum rod, 12.7mm (0.5in) dia, hard, 99.95% (metals basis)
CAS: 7440-25-7 | Ta | 180.95 g/mol
$241.85 - $3684.19
Chemical Identifiers
| CAS | 7440-25-7 |
|---|---|
| Molecular Formula | Ta |
| Molecular Weight (g/mol) | 180.95 |
| MDL Number | MFCD00011252 |
| InChI Key | GUVRBAGPIYLISA-UHFFFAOYSA-N |
| Synonym | tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod |
| PubChem CID | 23956 |
| ChEBI | CHEBI:33348 |
| IUPAC Name | tantalum |
| SMILES | [Ta] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AA10347BO
|
Thermo Scientific Chemicals
010347BO |
1 cm |
Each for $241.85
|
|
|||||
|
AA10347BQ
|
Thermo Scientific Chemicals
010347BQ |
5 cm |
Each for $868.53
|
|
|||||
|
AA10347BU
|
Thermo Scientific Chemicals
010347BU |
25 cm |
Each for $3,684.19
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-25-7 | |
| 180.95 | |
| GUVRBAGPIYLISA-UHFFFAOYSA-N | |
| 23956 | |
| tantalum |
| Ta | |
| MFCD00011252 | |
| tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
| CHEBI:33348 | |
| [Ta] |
Specifications
| 7440-25-7 | |
| Odorless | |
| Rod | |
| MFCD00011252 | |
| GUVRBAGPIYLISA-UHFFFAOYSA-N | |
| tantalum | |
| 23956 | |
| 99.95% | |
| (metals basis) |
| 1 cm | |
| ≈20.4g/cm | |
| Ta | |
| tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
| [Ta] | |
| 180.95 | |
| CHEBI:33348 | |
| Tantalum rod, 12.7mm (0.5 in.) dia., Hard, Temper: as drawn |
Safety and Handling
EINECSNumber : 231-135-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only