missing translation for 'onlineSavingsMsg'
Learn More

Tantalum powder, -325 mesh, Puratronic™, 99.97% (metals basis), Nb 50ppm

Catalog No. p-4408172
Change view
Click to view available options
Quantity:
2.5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1034588 2.5 g
AA1034514 25 g
AA1034522 100 g
AA1034536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1034588 Supplier Thermo Scientific Chemicals Supplier No. 01034588
Only null left

CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Color Black to Gray
Appearance Gray to black powder
Quantity 2.5 g
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Assay Percent Range 99.97% (Metals basis, Nb 50ppm)
Chemical Name or Material Tantalum
Hazard Category H228-H315-H319-H335
Precautionary Statement P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-135-5
RTECSNumber WW5505000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only