missing translation for 'onlineSavingsMsg'
Learn More

Tantalum powder, -325 mesh, 99.9% (metals basis), 99.6%

99.6% purity includes H, N, C, O | CAS: 7440-25-7 | Ta | 180.95 g/mol

Supplier:  Thermo Scientific Chemicals 00033722

 View more versions of this product

Catalog No. AA0033722


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Black to Gray
7440-25-7
100 g
Odorless
99.9% (Metals basis, 99.6% purity includes H, N, C, O)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
Tantalum
Gray to black powder
-325 Mesh
Powder
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.95
CHEBI:33348

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : DOT Class: 4.1, Packaging Group: II

EINECSNumber : 231-135-5

RTECSNumber : WW5505000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only