missing translation for 'onlineSavingsMsg'
Learn More

Tantalum powder, -22 mesh, Puratronic™, 99.95% (metals basis), Nb 100ppm

CAS: 7440-25-7 | Ta | 180.95 g/mol

$96.93 - $1062.71

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1075306
View Documents
Thermo Scientific Chemicals
01075306
5 g
Each for $96.93
Only null left
 
AA1075314
View Documents
Thermo Scientific Chemicals
01075314
25 g
Each for $333.64
Only null left
 
AA1075322
View Documents
Thermo Scientific Chemicals
01075322
100 g
Each for $1,062.71
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]

Specifications

Black to Gray
Gray to black powder
-22 Mesh
Powder
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.95
CHEBI:33348
7440-25-7
5 g
Odorless
99.98% (Metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
Tantalum
Videos
Safety and Handling

Safety and Handling

P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H228-H315-H319-H335

DOTInformation : DOT Class: 4.1; Packaging Group: II

EINECSNumber : 231-135-5

RTECSNumber : WW5505000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only