missing translation for 'onlineSavingsMsg'
Learn More

Tantalum gauze, 30 mesh woven from 0.076mm (0.003in) dia wire

Catalog No. p-7059102
Change view
Click to view available options
Quantity:
125 x 125 mm
250 x 250 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43965GN 125 x 125 mm
AA43965GY 250 x 250 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43965GN Supplier Thermo Scientific Chemicals Supplier No. 043965GN
Only null left

CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Quantity 125 x 125 mm
Mesh Size 30 Mesh
Form Wire Cloth
Chemical Name or Material Tantalum gauze, 30 mesh woven from 0.076mm (0.003 in.) dia. wire
EINECSNumber 231-135-5
TSCA Yes

RUO – Research Use Only