missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave, Thermo Scientific Chemicals
$264.79 - $264.79
Chemical Identifiers
| CAS | 7440-25-7 |
|---|---|
| Molecular Formula | Ta |
| Molecular Weight (g/mol) | 180.95 |
| MDL Number | MFCD00011252 |
| InChI Key | GUVRBAGPIYLISA-UHFFFAOYSA-N |
| Synonym | tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod |
| PubChem CID | 23956 |
| ChEBI | CHEBI:33348 |
| IUPAC Name | tantalum |
| SMILES | [Ta] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 7440-25-7 | |
| 180.95 | |
| GUVRBAGPIYLISA-UHFFFAOYSA-N | |
| 23956 | |
| tantalum |
| Ta | |
| MFCD00011252 | |
| tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
| CHEBI:33348 | |
| [Ta] |
Specifications
| 7440-25-7 | |
| 100 Mesh | |
| Wire Cloth | |
| MFCD00011252 | |
| GUVRBAGPIYLISA-UHFFFAOYSA-N | |
| tantalum | |
| 23956 | |
| Tantalum gauze, 100 mesh woven from 0.08mm (0.003 in.) dia. wire, twilled weave |
| 75 x 75 mm | |
| Odorless | |
| Ta | |
| tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod | |
| [Ta] | |
| 180.95 | |
| CHEBI:33348 |
Safety and Handling
EINECSNumber : 231-13505
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only