missing translation for 'onlineSavingsMsg'
Learn More

Tantalum foil, 1.5mm (0.06in) thick, 99.95% (metals basis)

CAS: 7440-25-7 | Ta | 180.95 g/mol

Supplier:  Thermo Scientific Chemicals 042115FI

 View more versions of this product

Catalog No. AA42115FI

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Tantalum foil
7440-25-7
Odorless
Foil
Ta
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
[Ta]
180.95
CHEBI:33348
1.5mm (0.06 in.) thick
50 x 50 mm
≈15.6g/25x25mm
(metals basis)
MFCD00011252
GUVRBAGPIYLISA-UHFFFAOYSA-N
tantalum
23956
99.95%

Chemical Identifiers

7440-25-7
180.95
GUVRBAGPIYLISA-UHFFFAOYSA-N
23956
tantalum
Ta
MFCD00011252
tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
CHEBI:33348
[Ta]
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 231-135-5

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only