missing translation for 'onlineSavingsMsg'
Learn More

Tantalum foil, 0.5mm (0.02in) thick, annealed, 99.95% (metals basis excluding Nb)

Catalog No. p-4408170
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
200 x 200 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA10355FF 25 x 25 mm
AA10355FI 50 x 50 mm
AA10355GH 100 x 100 mm
AA10355GW 200 x 200 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA10355FF Supplier Thermo Scientific Chemicals Supplier No. 010355FF
Only null left

Form: approx. 5.19g/25x25mm | CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Color Gray to Silver
Appearance Gray to silver foil
Quantity 25 x 25 mm
Odor Odorless
Form Foil
Assay Percent Range 99.95% (Metals basis excluding Nb)
Chemical Name or Material Tantalum, Annealed
EINECSNumber 231-135-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only