missing translation for 'onlineSavingsMsg'
Learn More

Tantalum foil, 0.025mm (0.001in) thick, hard, 99.95% (metals basis)

Catalog No. p-4408171
Change view
Click to view available options
Quantity:
100 x 150 mm
150 x 2000 mm
150 x 500 mm
Catalog No. Quantity
AA10351GJ 100 x 150 mm
AA10351IV 150 x 500 mm
AA10351IX 150 x 2000 mm
3 options

Catalog No. AA10351GJ

Supplier: Thermo Scientific Chemicals 010351GJ

Only null left

Form: approx. 6.3g/100x150mm; Temper: as rolled | CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Color Gray to Silver
Appearance Gray to silver foil
Density 16.69g/cm3
Quantity 100 x 150 mm
Odor Odorless
Form Foil
Assay Percent Range 99.95% (Metals basis)
Chemical Name or Material Tantalum, Hard
EINECSNumber 231-135-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only