missing translation for 'onlineSavingsMsg'
Learn More

Tantalum foil, 0.025mm (0.001in) thick, 99.997% (metals basis)

Catalog No. AA00335FQ
Change view
Click to view available options
Quantity:
25 x 25 mm
25 x 100 mm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA00335FQ 25 x 100 mm
AA00335FF 25 x 25 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA00335FQ Supplier Thermo Scientific Chemicals Supplier No. 000335FQ

missing translation for 'validMainframeMsg'

CAS: 7440-25-7 | Ta | 180.95 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-25-7
Molecular Formula Ta
Molecular Weight (g/mol) 180.95
MDL Number MFCD00011252
InChI Key GUVRBAGPIYLISA-UHFFFAOYSA-N
Synonym tantalum, elemental, hydride, tantalum-181, tantalum, metal and oxide dust, tantal, tantalum, metal, powder, foil, wire, rod
PubChem CID 23956
ChEBI CHEBI:33348
IUPAC Name tantalum
SMILES [Ta]

Specifications

Quantity 25 x 100 mm
Odor Odorless
Weight ≈0.26g/25x25mm
Form Foil
Percent Purity 99.997%
Assay (metals basis)
Chemical Name or Material Tantalum foil, 0.025mm (0.001 in.) thick
EINECSNumber 231-135-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only