missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tannic Acid Solution, 25% w/v, Reagents
Specifications
Specifications
| Chemical Name or Material | Tannic Acid Solution, 25% w/v |
| CAS | 7732-18-5, 1401-55-4, 531-32-1 |
| Boiling Point | 100°C |
| Color | Dark Brown |
| Quantity | 4 L |
| Packaging | Glass Bottle |
| Physical Form | Liquid |
| Specific Gravity | 1.1613 |
| Synonym | Gallotannic Acid, Glycerite |
| Percent Purity | 25% |
| Show More |