missing translation for 'onlineSavingsMsg'
Learn More

Tannic Acid, Reagents

Catalog No. p-200020896
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337559 100 g
01337558 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337559 Supplier Labchem Inc Supplier No. C2116000100C2
Only null left

Tannic Acid, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.21
InChI Key LRBQNJMCXXYXIU-UHFFFAOYNA-N
Synonym Gallotannic Acid, Glycerite
IUPAC Name 2,3-dihydroxy-5-[({3,4,5,6-tetrakis[3,4-dihydroxy-5-(3,4,5-trihydroxybenzoyloxy)benzoyloxy]oxan-2-yl}methoxy)carbonyl]phenyl 3,4,5-trihydroxybenzoate
SMILES OC1=CC(=CC(O)=C1O)C(=O)OC1=CC(=CC(O)=C1O)C(=O)OCC1OC(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C(OC(=O)C2=CC(O)=C(O)C(OC(=O)C3=CC(O)=C(O)C(O)=C3)=C2)C1OC(=O)C1=CC(O)=C(O)C(OC(=O)C2=CC(O)=C(O)C(O)=C2)=C1

Specifications

Color Off-White to Light Brown
Grade Reagent
Quantity 100 g
Packaging Glass Bottle
Physical Form Solid
Specific Gravity 2.807
Chemical Name or Material Tannic Acid