missing translation for 'onlineSavingsMsg'
Learn More

Tannic Acid, Honeywell Fluka™

Catalog No. p-7071832
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
25 kg
6 x 1 kg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6002020 100 g
6002024 500 g
6002021 1 kg
60044932 6 x 1 kg
6002023 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 6002020 Supplier Honeywell Chemicals Supplier No. 16201100G
Only null left

Puriss., meets analytical specification of USP, powder

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.206
MDL Number MFCD00066397
InChI Key LRBQNJMCXXYXIU-JPZKJCTESA-N
Synonym tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
PubChem CID 134129492
SMILES C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(

Specifications

UN Number NONH for all modes of transport
Beilstein 8186396
Solubility Information Soluble in acetone, alcohol, water 1gm/0.35mL, glycerol 1gm/1mL (warm). Insoluble in carbon disulfide, chloroform, diethyl ether,carbon tetrachloride, petroleum ether,benzene, hexane. Slightly soluble in dehydrated alcohol.
Formula Weight 1701.2g/mol
Grade puriss.
Quantity 100 g
Flash Point 199°C (390.2°F)
Ignition Point 980°F
Packaging Plastic Bottle
Physical Form Powder
Residue after Ignition ≤0.1% (as SO4)
Chemical Name or Material Tannic Acid, Meets analytical specification of USP
Show More Show Less
Precautionary Statement H412
EINECSNumber 215-753-2
RTECSNumber WW5075000