missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tannic acid, ACS reagent
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 218.0°C |
| Color | Brown |
| Quantity | 500 g |
| Flash Point | 198°C |
| Infrared Spectrum | Authentic |
| Loss on Drying | 12.0% max. (105°C) |
| Packaging | Plastic bottle |
| Physical Form | Powder |
| Residue after Ignition | 0.5% max. |
| Merck Index | 15, 9184 |
| Show More |
Safety and Handling
| Hazard Category | LIGHT SENSITIVE |
| Hazard Statement | AIR SENSITIVE |
| EINECSNumber | 215-753-2 |
| RTECSNumber | PY7955000 |
| TSCA | TSCA |
RUO – Research Use Only