missing translation for 'onlineSavingsMsg'
Learn More

Tannic acid, 95%

Catalog No. p-3776552
Change view
Click to view available options
Quantity:
5 g
500 g
1 kg
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC202420050 5 g Glass bottle
AC202425000 500 g Plastic bottle
AC202420010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC202420050 Supplier Thermo Scientific Chemicals Supplier No. 202420050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1401-55-4 | C76H52O46 | 1701.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1401-55-4
Molecular Formula C76H52O46
Molecular Weight (g/mol) 1701.206
MDL Number MFCD00066397
InChI Key LRBQNJMCXXYXIU-JPZKJCTESA-N
Synonym tannins, gallotannic acid, quebracho extract, acide tannique, d'acide tannique, tannic acid usp:jan, unii-28f9e0djy6
PubChem CID 134129492
SMILES C1=C(C=C(C(=C1O)O)O)C(=O)OC2=CC(=CC(=C2O)O)C(=O)OCC3C(C(C(C(O3)OC(=O)C4=CC(=C(C(=C4)OC(=O)C5=CC(=C(C(=C5)O)O)O)O)O)OC(=O)C6=CC(=C(C(=C6)OC(=O)C7=CC(=C(C(=C7)O)O)O)O)O)OC(=O)C8=CC(=C(C(=C8)OC(=O)C9=CC(=C(C(=C9)O)O)O)O)O)OC(=O)C1=CC(=C(C(=C1)OC(=O)C1=CC(=C(

Specifications

Melting Point 218.0°C
Color White to Yellow
Density 198g/mL
Quantity 5 g
Flash Point 198°C
Infrared Spectrum Authentic
Loss on Drying 12.0% max.
Packaging Glass bottle
Physical Form Beads
Assay Percent Range 95%
Merck Index 15, 9184
Solubility Information Solubility in water: 250g/L (20°C). Other solubilities: soluble in glycerol, ethanol and acetone;, practically insoluble in benzene, chcl3, diethyl e, ther, petroleum ether, cs2, ccl4, hexane;, slightly soluble in anhydrous ethanol.
Formula Weight 1701.23
Percent Purity 95%
Chemical Name or Material Tannic acid
Show More Show Less

RUO – Research Use Only