Learn More
Corning™ Tank Liners
Designed to fit standardized cylindrical tanks manufactured by Nalge™/Nunc™ and other brands
Supplier:  Corning™ 9130020
 View more versions of this product
Catalog No. MT9130020
Only null left
Description
Description
  • Made from low-density polyethylene (LDPE)
  • Eliminate costs associated with cleaning and validation of tanks
  • Decrease labor costs and increase turnaround time
  • Eliminate risks associated with cross contamination
  • Help extend life of reuseable tanks, resulting in reduced capital expenditures
  • Gamma irradiated, individually packaged
  • Choice of gusseted and non-gusseted available
  • Available in many sizes
Specifications
Specifications
130 L
34.3 gal.
45.72 cm
48 in. x 18 in.
LDPE
18 in.
Cylindrical
Non-Gusseted Tank Liner
Product Suggestions
Product Suggestions