missing translation for 'onlineSavingsMsg'
Learn More

Tank Liner, Aegis™ 5-14, top and bottom drain

Catalog No. SH3130601
Change view
Click to view available options
Capacity (Metric):
50 L
100 L
200 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
SH3130601 50 L
SH3130602 100 L
SH3130603 200 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. SH3130601 Supplier Thermo Fisher Scientific Supplier No. SH31306.01

Use Thermo Scientific Tank Liners for non-sterile fluid handling operations such as waste collection or powdered media or buffer hydration processes. Tank Liners are constructed with our Thermo Scientific Aegis 5-14 single web, multi-layered cast film manufactured in a cGMP clean room facility.

Features include:
• Film optimized for flexibility and durability: superior stress resistance, puncture resistance and physical barrier properties to ensure stability of contents
• Gamma-irradiation sterilized
• Squared tube design for best fit and reliability
• Space-efficient, ergonomic alternative to rigid walled carboys and steel tanks
• Customizable via the industry's largest library of qualified components

Recommended for:
• Harvest—collection and storage of bulk non-sterile process liquids
• Collect—non-sterile process liquids for further processing
• Hydrate—powdered buffers, media, and media supplements

Specifications

Capacity (Metric) 50 L
Quantity 50 L
Tubing Style TPE