missing translation for 'onlineSavingsMsg'
Learn More

Tamoxifen citrate

Catalog No. p-7041987
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6095503 1 g
AAJ6095506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ6095503 Supplier Thermo Scientific Chemicals Supplier No. J6095503

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Inhibitor of protein kinase C and induces apoptosis in cancer cells | CAS: 54965-24-1 | C32H37NO8 | 563.65 g/mol

Tamoxifen citrate is used as an antiestrogen with potent anti-cancer and PKC inhibitory activities. Tamoxifen Citrate functions as an estrogen receptor antagonist in breast tissue. Tamoxifen Citrate is an agonist towards this receptor in tissues of the endometrium. Tamoxifen Citrate inhibits DNA synthesis and transcription by recruiting co-repressors, which stop estrogen from interacting with genes. When combined with PAX2, Tamoxifen Citrate can inhibit the proliferation of ERBB2. It is also a high affinity activator of the GPR30 receptor. Tamoxifen Citrate is an activator of Estrogen Receptor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tamoxifen citrate is used as an antiestrogen with potent anti-cancer and PKC inhibitory activities. Tamoxifen Citrate functions as an estrogen receptor antagonist in breast tissue. Tamoxifen Citrate is an agonist towards this receptor in tissues of the endometrium. Tamoxifen Citrate inhibits DNA synthesis and transcription by recruiting co-repressors, which stop estrogen from interacting with genes. When combined with PAX2, Tamoxifen Citrate can inhibit the proliferation of ERBB2. It is also a high affinity activator of the GPR30 receptor. Tamoxifen Citrate is an activator of Estrogen Receptor.

Solubility
Soluble in methanol (50 mg/ml with heat) or ethanol (10 mg/ml with sonication); very slightly soluble in water (0.3 mg/L @ 20°C; the pH is approximately 3.0-3.5), 0.02 N HCl (0.2 mg/ml @ 37°C)4, acetone ,chloroform,and DMSO(100mM)

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: 2 - 8°C. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 54965-24-1
Molecular Formula C32H37NO8
Molecular Weight (g/mol) 563.65
MDL Number MFCD00058321
InChI Key FQZYTYWMLGAPFJ-OQKDUQJOSA-N
Synonym tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen
PubChem CID 2733525
ChEBI CHEBI:9397
IUPAC Name (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1

Specifications

Color White
Quantity 1 g
Sensitivity Hygroscopic; Light sensitive
Merck Index 14,9048
Solubility Information Soluble in methanol (50mg/ml with heat) or ethanol (10mg/ml with sonication); very slightly soluble in water (0.3mg/L @ 20°C; the pH is approximately 3.0-3.5),0.02 N HCl (0.2mg/ml @ 37°C)4,acetone ,chloroform,and DMSO(100mM)
Formula Weight 563.65
Physical Form Powder
Chemical Name or Material Tamoxifen citrate
Hazard Category H302-H350
Hazard Statement GHS H Statement
H350-H360-H362-H302
May cause cancer.
May damage fertility or the unborn child.
May cause harm to breast-fed children.
Harmful if swallowed.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 259-415-2
RTECSNumber KH2387000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only