missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tamoxifen Citrate Salt, MP Biomedicals™
$86.68 - $86.68
Chemical Identifiers
| CAS | 54965-24-1 |
|---|---|
| Molecular Formula | C32H37NO8 |
| Molecular Weight (g/mol) | 563.65 |
| MDL Number | MFCD00058321 |
| InChI Key | FQZYTYWMLGAPFJ-OQKDUQJOSA-N |
| Synonym | tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen |
| PubChem CID | 2733525 |
| ChEBI | CHEBI:9397 |
| IUPAC Name | (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid |
| SMILES | OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15673980
|
MP Biomedicals Inc
0215673980 |
100 mg |
Each for $86.68
|
|
|||||
Description
Molecular biologyChemical Identifiers
| 54965-24-1 | |
| 563.65 | |
| FQZYTYWMLGAPFJ-OQKDUQJOSA-N | |
| 2733525 | |
| (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid |
| C32H37NO8 | |
| MFCD00058321 | |
| tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen | |
| CHEBI:9397 | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 |
Specifications
| 54965-24-1 | |
| C32H37NO8 | |
| 100 mg | |
| FQZYTYWMLGAPFJ-OQKDUQJOSA-N | |
| (2-{4-[(1Z)-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine; 2-hydroxypropane-1,2,3-tricarboxylic acid | |
| 2733525 | |
| 563.65 | |
| Tamoxifen Citrate Salt |
| White | |
| MFCD00058321 | |
| tamoxifen citrate, istubal, zitazonium, unii-7frv7310n6, i.c.i. 46474 citrate, tamoxifen citrate salt, farmifeno, ginarsan, jenoxifen | |
| OC(=O)CC(O)(CC(O)=O)C(O)=O.CC\C(=C(/C1=CC=CC=C1)C1=CC=C(OCCN(C)C)C=C1)C1=CC=CC=C1 | |
| 563.65 | |
| CHEBI:9397 | |
| Solid |
Safety and Handling
EINECSNumber : 259-415-2
storageNote1 : Store desiccated. Protect from light
Recommended Storage : Store at +4°C