Learn More
Stoelting™ Tail Access Rodent Restrainer
Stoelting™ Tail Access Rodent Restrainer is ideal for injections, blood sampling, or blood pressure monitoring
$420.51 - $447.85
Specifications
For Use With (Application) Neuroscience
Animal Rodent
View More Specs
Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
10000749
View Documents Promotion Details
Stoelting™
51337
30 to 125 g
Each for $447.85
Only null left
10000750
View Documents Promotion Details
Stoelting™
STG51338
15 to 30 g
Each for $420.51
Only null left
Description
Description
  • Manufactured from clear materials thus providing complete visual awareness of the animal
  • Animal's head is pushed back by the moveable disk so that the tail projects out the hole in the acrylic plate
  • Offered in four sizes to accomodate indicated rodent weight.
Specifications
Specifications
Neuroscience
Rodent