Learn More
High-Tech Conversions Tacky Traxx™ Adhesive Cleanroom Mats
Tacky Traxx™ Adhesive Cleanroom Mats are designed to remove particulates created by foot and cart traffic.
Supplier:  High-Tech Conversions TT366034B
 View more versions of this product
Catalog No. 501047000
Only null left
Explore available promotions
Description
Description
  • Numbered corner tabs for easy tracking of remaining sheets
  • Smooth surface is not damaged by shoes, booties, or cart wheels
  • Peel-off sheets eliminate the need for messy cleaning, making it easy to maintain a clean surface
  • Ideal for cleanrooms, laboratories, construction, manufacturing, maintenance, installation service, and residential use
Specifications
Specifications
Single-use
Entrances in Particle-Controlled Environments
30 Sheets/Mat
Polyethylene
Peel-off sheets
Blue
91.5 cm
60 in.
Cleanroom Tacky Entrance Mat
Polyethylene Adhesive Film
36 x 60 in.
36 in.
152.5 cm
Product Suggestions
Product Suggestions