missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TABS, 99%

Chemical Identifiers

CAS 54960-65-5
Synonym N-Tris(hydroxymethyl)methyl-4-aminobutanesulfonic acid
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC428080250
SDS View Documents
Thermo Scientific™
428080250
25 g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC428081000
SDS View Documents
Thermo Scientific™
428081000
100 g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Biological buffer
Specifications

Chemical Identifiers

54960-65-5
N-Tris(hydroxymethyl)methyl-4-aminobutanesulfonic acid

Specifications

257.301
Glass bottle
N-Tris(hydroxymethyl)methyl-4-aminobutanesulfonic acid
≥98.5% (on anhydrous basis)
25 g
54960-65-5
Crystalline Powder
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only