Learn More
SOMNI Scientific™ Tabletop Rebreathing Small Animal Anesthesia Machine
Catalog No. 04355315
Encompass
Change view
Click to view available options
Animal:
Large Animal
Catalog No. Animal
04355315 Large Animal
1 options
Catalog No. 04355315 Supplier SOMNI Scientific™ Supplier No. RM17008
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The SOMNI Tabletop Rebreathing Small Animal Anesthesia Machine is made of high-quality materials and has a small footprint. The compact design provides exceptional stability and the fresh gas outlet conveniently routes the anesthesia from rebreathing to nonrebreathing
  • High quality limited maintenance design
  • Pressure manometer center mounted for immediate visibility
  • Toggle switch: rebreathing to nonrebreathing, circuits stay in place
  • Popoff valve releases at 2 cm H2O while maintaining passive volume
  • Small footprint, easy assembly
  • Powder coated and anodized frame for easy cleaning
Specifications
For Use With (Application) Animal Inhalant Anesthesia
Animal Large Animal
Type Anesthesia Machine