The resDetect T7 RNA Polymerase ELISA Kit is a complete sandwich ELISA for sensitive, quantitative detection of T7 RNA polymerase residues in cell culture supernatants. Designed for residue monitoring during mRNA production, the kit supplies pre-coated plates, standards, detection antibodies, streptavidin-HRP, buffers, substrate, and stop solution in a 96-test format for research use.
- 96-test format (8 x 12 strips) compatible with standard microplate readers.
- Sensitivity less than 0.78 ng/mL; standard curve range 0.78-50 ng/mL.
- Includes pre-coated microplate, standard, biotin-antibody, streptavidin-HRP, wash and dilution buffers, substrate, and stop solution.
- Sample type: cell culture supernatants; sample volume 100 L.
- Assay time approximately 3 h 20 min.
- Store unopened kit at 2-8 C; unopened stability 12 months.
- For research use only (RUO).