Learn More
New England Biolab Ltd T7 Express Competent E. coli (High Efficiency) 20 x 0.05 ml/tube
Catalog No. 50994938
Encompass
High efficiency chemically competent E. coli cells suitable for T7 protein expression.
Catalog No. 50994938 Supplier New England Biolab Ltd Supplier No. C2566H
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
High efficiency chemically competent E. coli cells suitable for T7 protein expression.