Learn More
New England Biolab Ltd T7 Endonuclease I 250 units
Catalog No. 50995225
Encompass
T7 Endonuclease I recognizes and cleaves non-perfectly matched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA and more slowly, nicked double-stranded DNA. The cleavage site is at the first, second or third phosphodiester bond that is 5' to the mismatch. The protein is the product of T7 gene 3.
Catalog No. 50995225 Supplier New England Biolab Ltd Supplier No. M0302S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
T7 Endonuclease I recognizes and cleaves non-perfectly matched DNA, cruciform DNA structures, Holliday structures or junctions, heteroduplex DNA and more slowly, nicked double-stranded DNA. The cleavage site is at the first, second or third phosphodiester bond that is 5' to the mismatch. The protein is the product of T7 gene 3.