Learn More
New England Biolab Ltd T5 Exonuclease 5000 units
Catalog No. 502024043
Encompass
T5 Exonuclease degrades DNA in the 5 to 3 direction. T5 Exonuclease is able to initiate nucleotide removal from the 5 termini or at gaps and nicks of linear or circular dsDNA. However, the enzyme does not degrade supercoiled dsDNA. T5 Exonuclease also has ssDNA endonuclease activity.
Catalog No. 502024043 Supplier New England Biolab Ltd Supplier No. M0663L
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
T5 Exonuclease degrades DNA in the 5 to 3 direction. T5 Exonuclease is able to initiate nucleotide removal from the 5 termini or at gaps and nicks of linear or circular dsDNA. However, the enzyme does not degrade supercoiled dsDNA. T5 Exonuclease also has ssDNA endonuclease activity.