missing translation for 'onlineSavingsMsg'
Learn More

T4 UvsX DNA Recombinase, Glycerol Free, G-Biosciences

Catalog No. S000287
Change view
Click to view available options
Quantity:
100 μg
1000 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S000287 1000 μg
S000286 500 μg
S000284 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S000287 Supplier G-Biosciences Supplier No. 7862135

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

T4 UvsX DNA recombinase mediates DNA strand exchange between homologous chromosomes.

The protein forms a right-handed nucleoprotein complex on single ssDNA called the presynaptic filament that can search for homology in duplex DNA and pair the recombining DNA molecules to form a DNA joint molecule. This process is aided by ssDNA binding proteins and recombination mediators. The filament is then elongated by DNA polymerase with the newly synthesized strand displacing the old strand. The newly synthesized duplex DNA can then act as a template for the next cycle to achieve exponential amplification of dsDNA

Specifications

Detection Method Gel Electrophoresis
Form Liquid
GC-Rich PCR Performance High
PCR Method End-point PCR
Reaction Speed 20 min.
Label or Dye Biotin, FAM, TAMRA
Product Type Uvsx recombinase
Quantity 1000 μg
For Use With (Application) Microbial Detection, Pathogen Detection, Isothermal Amplification