Learn More
New England Biolab Ltd T4 DNA Polymerase 150 units
Catalog No. 50811608
Encompass
T4 DNA Polymerase catalyzes the synthesis of DNA in the 5' to 3' direction and requires the presence of template and primer. This enzyme has a 3' to 5' exonuclease activity which is much more active than that found in DNA Polymerase I (E. coli). Unlike E. coli DNA Polymerase I, T4 DNA Polymerase does not have a 5' to 3' exonuclease function.
Catalog No. 50811608 Supplier New England Biolab Ltd Supplier No. M0203S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
T4 DNA Polymerase catalyzes the synthesis of DNA in the 5' to 3' direction and requires the presence of template and primer. This enzyme has a 3' to 5' exonuclease activity which is much more active than that found in DNA Polymerase I (E. coli). Unlike E. coli DNA Polymerase I, T4 DNA Polymerase does not have a 5' to 3' exonuclease function.