Learn More
Thermo Scientific™ T-valve Assembly for LN2 Supply Tank
Catalog No. TF4000290
Change view
Click to view available options
For Use With (Equipment):
CryoPlus 1,2,3,4
Catalog No. For Use With (Equipment)
TF4000290 CryoPlus 1,2,3,4
1 options
Catalog No. TF4000290 Supplier Thermo Scientific™ Supplier No. 4000290
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Connect two hoses to a LN2 tank with this T-valve assembly, which includes a manual valve and T-fitting.
Connect two hoses to a LN2 tank with this Thermo Scientific™ T-valve assembly for LN2 supply tank, which includes a manual valve and T-fitting.
Specifications
Description Manual valve and T fitting
Type Manual Valve and T-Fitting
For Use With (Equipment) CryoPlus 1,2,3,4
Defrost Manual