Learn More
Promega T Cell Activation Bioassay (NFAT), 1 each
The T Cell Activation Bioassays are bioluminescent cell-based assays that overcome the limitations of existing assays and can be used for the discovery and development of novel biologics such as bispecific antibodies and CAR-T cell therapies.
Supplier:  Promega J1621
Encompass
Catalog No. PRJ1621
Only null left
Description
Description
  • Measure Stability and Precision of Biologics Designed to Affect T Cell Responses
  • Prequalified according to ICH guidelines
  • Amenable to high−throughput formats
  • No cell culture required
Specifications
Specifications
COMPONENTS AT DIFFERENT TEMPS
1 Each
Product Suggestions
Product Suggestions