missing translation for 'onlineSavingsMsg'
Learn More

Lovibond™ T-CAL™ Turbidity Standard, 20 NTU

Catalog No. 15398370
Change view
Click to view available options
Volume (Metric):
1000 mL/1 L
125 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
15398370 1000 mL/1 L
15398368 125 mL
15398369 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15398370 Supplier Lovibond™ Supplier No. 48012399

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

T-CAL™ Turbidity Standards are stabilized Formazin standards prepared in ready-to-use concentrations to ensure accurate turbidity measurements. Eliminate the opportunity for inaccurate turbidimeter calibrations resulting from the improper dilution or handling of Formazin stock solution.

  • Save time and eliminate errors - no need for dilution
  • EPA approved primary turbidity standard
  • Long shelf life
  • All concentrations of T-CAL™ Turbidity Standards are verified to meet tight quality control specifications, are EPA and ISO compliant for reporting purposes and guaranteed to save you time. As a primary standard, T-CAL™ Turbidity Standards can be used to calibrate or verify any manufacturer's turbidimeter.

Specifications

For Use With (Equipment) Turbidimeter
Calibration Type Primary
Form Liquid
For Use With (Application) Calibration or Verification of a Turbidimeter
Turbidity 20 NTU
Certifications/Compliance EN ISO 7027, USEPA 180.1 and APHA 2130 Part B.
Volume (Metric) 1000 mL/1 L
Quantity 1 Ea.
Packaging Type Amber Plastic Bottles